Stampa…
2-Amino-4-methyl-3-nitropyridine ≥98.0% (by GC, titration analysis)
Fornitore: TCI EUROPE
Attenzione
Synonyms: 4-Methyl-3-nitro-2-pyridylamine, 4-Methyl-3-nitropyridin-2-amine, 2-Amino-3-nitro-4-picoline
A1888-1G A1888-5G
TCIAA1888-1GEA 16 EUR
TCIAA1888-1G TCIAA1888-5G
2-Amino-4-methyl-3-nitropyridine ≥98.0% (by GC, titration analysis)
2-Amino-4-methyl-3-nitropyridine
Formula: C₆H₇N₃O₂
Peso molecolare: 153,14 g/mol
Punto di fusione: 140 °C
MDL Number: MFCD00006315
CAS Number: 6635-86-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=963294086640316bd8a62b0869f5ca183bedcee8&vid=963294086640316bd8a62b0869f5ca183bedcee8&prid=web&clid=SAPDAM
Visualizza i codici
Appearance Light orange to Yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 136.0 to 140.0 deg-C