Stampa…

2-Aminofluorene ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI
Attenzione

Synonyms: 9H-Fluoren-2-amine, 2-Fluorenamine, Fluoren-2-ylamine

A0621-25G A0621-5G
TCIAA0621-25GEA 139 EUR
TCIAA0621-25G TCIAA0621-5G
2-Aminofluorene ≥98.0% (by HPLC, titration analysis)
2-Aminofluorene
Formula: C₁₃H₁₁N
Peso molecolare: 181.24 g/mol
Punto di ebollizione: 327
Densità: 1.281
MDL Number: MFCD00001125
CAS Number: 153-78-6
EINECS: 205-817-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d61875f97a9aeb6da411624ea503ae13c98d10b4&vid=d61875f97a9aeb6da411624ea503ae13c98d10b4&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 126.0 to 129.0 deg-C
Solubility in hot Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR