3-Iodopropionic acid ≥98.0% (by GC, titration analysis)
Fornitore: TCI EUROPE
|
Pericolo
|
Synonyms:
3-Iodopropanoic acid, 3-Iodopropionicacid
I0071-25G
I0071-5G
I0071-100G
TCIAI0071-25GEA
61
EUR
TCIAI0071-25G
TCIAI0071-5G
TCIAI0071-100G
3-Iodopropionic acid ≥98.0% (by GC, titration analysis)
3-Iodopropionic acid
Formula:
C₃H₅IO₂ Peso molecolare: 199,98 g/mol Punto di ebollizione: 260 °C (1013 hPa) Punto di fusione: 83 °C |
MDL Number:
MFCD00002765 CAS Number: 141-76-4 EINECS: 205-499-0 UN: 3261 ADR: 8,III |
Risultati del test di specificazione
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 82.0 to 86.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...