Stampa…

2-Iodo-4-(trifluoromethyl)aniline ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 4-Amino-3-iodobenzotrifluoride, 2-Iodo-α,α,α-trifluoro-p-toluidine, 4-Amino-3-iodo-α,α,α-trifluorotoluene

I0794-25G I0794-5G
TCIAI0794-25GEA 171 EUR
TCIAI0794-25G TCIAI0794-5G
2-Iodo-4-(trifluoromethyl)aniline ≥98.0% (by GC)
2-Iodo-4-(trifluoromethyl)aniline
Formula: C₇H₅F₃IN
Peso molecolare: 287,02 g/mol
MDL Number: MFCD02683097
CAS Number: 163444-17-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dd17f1ffef0be689f1dd6d1893b3b235e6a64736&vid=dd17f1ffef0be689f1dd6d1893b3b235e6a64736&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Light yellow to Brown powder to crystal
Purity (GC) min. 98.0 %
Melting point 50.0 to 54.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR