Stampa…

2-Fluoro-5-iodobenzoic acid ≥98.0% (by titrimetric analysis)

Fornitore: TCI EUROPE
Attenzione

F0786-1G F0786-5G
TCIAF0786-1GEA 38 EUR
TCIAF0786-1G TCIAF0786-5G
2-Fluoro-5-iodobenzoic acid ≥98.0% (by titrimetric analysis)
2-Fluoro-5-iodobenzoic acid
Formula: C₇H₄FIO₂
Peso molecolare: 266,01 g/mol
MDL Number: MFCD03094517
CAS Number: 124700-41-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4bba761334d240bb4607c7e94b5eff17d8a26494&vid=4bba761334d240bb4607c7e94b5eff17d8a26494&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Brown powder to crystal
Purity (GC) min. 96.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 163.0 to 167.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR