(α,α,α-Trifluoro-2-tolyl)acetic acid ≥98.0% (by GC, titration analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
2-(Trifluoromethyl)phenylacetic acid, (2-(Trifluoromethyl)phenyl)acetic acid
T1661-1G
T1661-25G
T1661-5G
TCIAT1661-1GEA
16
EUR
TCIAT1661-1G
TCIAT1661-25G
TCIAT1661-5G
(α,α,α-Trifluoro-2-tolyl)acetic acid ≥98.0% (by GC, titration analysis)
(α,α,α-Trifluoro-2-tolyl)acetic acid
Formula:
F₃CC₆H₄CH₂COOH Peso molecolare: 204,15 g/mol Punto di ebollizione: 272 °C (1013 hPa) Punto di fusione: 98…100 °C |
MDL Number:
MFCD00004327 CAS Number: 3038-48-0 EINECS: 221-240-4 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 100.0 to 104.0 deg-C |
Solubility in Methanol | very faint turbidity |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...