Stampa…

2,3,5-Trifluorobenzoic acid ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

T2385-1G T2385-5G
TCIAT2385-1GEA 87 EUR
TCIAT2385-1G TCIAT2385-5G
2,3,5-Trifluorobenzoic acid ≥98.0% (by GC, titration analysis)
2,3,5-Trifluorobenzoic acid
Formula: C₇H₃F₃O₂
Peso molecolare: 176,09 g/mol
MDL Number: MFCD00061200
CAS Number: 654-87-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5549735fc90596dc814a07fab04ae9cc32291157&vid=5549735fc90596dc814a07fab04ae9cc32291157&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 107.0 to 111.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR