2,3,4,5-Tetrafluorobenzoyl chloride ≥97.0% (by GC, titration analysis)
Fornitore: TCI EUROPE
|
Pericolo
|
Synonyms:
2,3,4,5-tetrafluorobenzoylchloride
T1855-25G
T1855-5G
TCIAT1855-25GEA
43
EUR
TCIAT1855-25G
TCIAT1855-5G
2,3,4,5-Tetrafluorobenzoyl chloride ≥97.0% (by GC, titration analysis)
2,3,4,5-Tetrafluorobenzoyl chloride
Formula:
C₇HClF₄O Peso molecolare: 212,53 g/mol Punto di ebollizione: 53 - 54 °C (1013 hPa) Densità: 1,58 g/cm³ (20 °C) Flash Pt: 89°C Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00075164 CAS Number: 94695-48-4 UN: 3265 ADR: 8,III |
Risultati del test di specificazione
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 97.0 % |
Purity (Argentmetric Titration) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...