Stampa…

4-Fluorochalcone ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 4-Fluorobenzylidenacetophenone, 3-(4-Fluorophenyl)-1-phenylprop-2-en-1-one

F0459-25G F0459-5G
TCIAF0459-25GEA 91 EUR
TCIAF0459-25G TCIAF0459-5G
4-Fluorochalcone ≥98.0% (by GC)
4-Fluorochalcone
Formula: C₁₅H₁₁FO
Peso molecolare: 226,25 g/mol
MDL Number: MFCD00443467
CAS Number: 1608-51-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=df48743891ddd87e5b0f006af9e4debc83cf9df2&vid=df48743891ddd87e5b0f006af9e4debc83cf9df2&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Light orange to Yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 87.0 to 91.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR