Fluorotriphenylsilane ≥97.0% (by GC)
Fornitore: TCI
|
Pericolo
|
F0439-25G
F0439-5G
TCIAF0439-25GEA
195
EUR
TCIAF0439-25G
TCIAF0439-5G
Fluorotriphenylsilane ≥97.0% (by GC)
Fluorotriphenylsilane
Formula:
C₁₈H₁₅FSi Punto di ebollizione: 210°C Punto di fusione: 63 °C Densità: 1,212 g/cm³ (20 °C) |
MDL Number:
MFCD00017899 CAS Number: 379-50-0 EINECS: 206-832-2 UN: 3261 ADR: 8,II |
|
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 97.0 % |
Melting point | 62.0 to 66.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...