Pyridine-2,3-dicarboxylic acid ≥98.0% (by GC, titration analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
Quinolinic acid
P0550-25G
P0550-500G
TCIAP0550-25GEA
16
EUR
TCIAP0550-25G
TCIAP0550-500G
Pyridine-2,3-dicarboxylic acid ≥98.0% (by GC, titration analysis)
Pyridine-2,3-dicarboxylic acid
Formula:
C₇H₅NO₄ Peso molecolare: 167,12 g/mol Punto di ebollizione: 357 °C (1013 hPa) Punto di fusione: 188°C Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00006295 CAS Number: 89-00-9 EINECS: 201-874-8 Merck Index: 14,08073 |
Risultati del test di specificazione
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Solubility in hot Water | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...