Potassio D-gluconato ≥99.0% (by titrimetric analysis)
Fornitore: TCI EUROPE
Synonyms:
Potassium Gluconate, Gluconic Acid Potassium Salt, D-Gluconic acid potassium salt, Potassium (2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoate
G0040-25G
G0040-500G
TCIAG0040-25GEA
16
EUR
TCIAG0040-25G
TCIAG0040-500G
Potassio D-gluconato ≥99.0% (by titrimetric analysis)
Potassio D-gluconato
Formula:
HOH₂C(CH(OH))₄COOK Peso molecolare: 234,25 g/mol Punto di fusione: 174…176 °C Densità: 1,73 g/cm³ (20 °C) |
MDL Number:
MFCD00064211 CAS Number: 299-27-4 EINECS: 206-074-2 Merck Index: 14,04456 |
Risultati del test di specificazione
Appearance | White to Light yellow powder to crystal |
Purity (Nonaqueous Titration) | min. 99.0 % (calcd.on dried substance) |
Specific rotation [a]20/D | +10.5 to +15.0 deg (C=2, H2O) |
Solubility in Water | almost transparency |
Drying loss | max. 3.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...