Stampa…

trans-2-Hydroxycinnamic acid ≥98.0% (by GC, titration analysis)

Fornitore: TCI
Attenzione

Synonyms: (E)-3-(2-Hydroxyphenyl)acrylic acid, 2-Hydroxycinnamic acid, predominantly trans, trans-2-Hydroxycinnamic acid, trans-o-Coumaric acid, o-Coumaric acid

C0394-25G C0394-5G
TCIAC0394-25GEA 79 EUR
TCIAC0394-25G TCIAC0394-5G
trans-2-Hydroxycinnamic acid ≥98.0% (by GC, titration analysis)
trans-2-Hydroxycinnamic acid
Formula: C₉H₈O₃
Peso molecolare: 164.16 g/mol
Punto di ebollizione: 330
Punto di fusione: 217
MDL Number: MFCD00004379
CAS Number: 614-60-8
EINECS: 210-386-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8040c568a32de4da689ad88da669d961397045a3&vid=86b0604a76d1d2f16626e2fb4fe35aad6d0cb1b2&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR