Stampa…
4-Hydroxy-o-toluic acid ≥98.0% (by GC)
Fornitore: TCI EUROPE
Attenzione
Synonyms: 4-Hydroxy-2-methylbenzoic acid, 4-Hydroxy-o-toluic acid
H1311-25G H1311-5G
TCIAH1311-25GEA 153 EUR
TCIAH1311-25G TCIAH1311-5G
4-Hydroxy-o-toluic acid ≥98.0% (by GC)
4-Hydroxy-o-toluic acid
Formula: C₈H₈O₃
Peso molecolare: 152,15 g/mol
MDL Number: MFCD02182261
CAS Number: 578-39-2
EINECS: 209-422-1
Visualizza i codici
Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 97.0 %
Melting point 177.0 to 181.0 deg-C