2,3,4-Trimethoxycinnamic acid ≥98.0% (by GC, titration analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
3-(2,3,4-Trimethoxyphenyl)acrylic acid
T2016-25G
T2016-5G
TCIAT2016-25GEA
188
EUR
TCIAT2016-25G
TCIAT2016-5G
2,3,4-Trimethoxycinnamic acid ≥98.0% (by GC, titration analysis)
2,3,4-Trimethoxycinnamic acid
Formula:
C₁₂H₁₄O₅ Peso molecolare: 238,24 g/mol Punto di ebollizione: 367 °C (1013 hPa) Punto di fusione: 174…176 °C |
MDL Number:
MFCD00014376 CAS Number: 33130-03-9 EINECS: 251-388-5 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 170.0 to 175.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...