Stampa…

3-Hydroxy-4-methoxycinnamic acid ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: Isoferulic acid, 3-(3-Hydroxy-4-methoxyphenyl)acrylic acid

H0524-1G H0524-5G
TCIAH0524-1GEA 59 EUR
TCIAH0524-1G TCIAH0524-5G
3-Hydroxy-4-methoxycinnamic acid ≥98.0% (by HPLC, titration analysis)
3-Hydroxy-4-methoxycinnamic acid
Formula: C₁₀H₁₀O₄
Peso molecolare: 194,184 g/mol
MDL Number: MFCD00004391
CAS Number: 537-73-5
EINECS: 208-676-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=11b6f58bdd7490d89c3ab430f4ace1d7a3bdb514&vid=11b6f58bdd7490d89c3ab430f4ace1d7a3bdb514&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR