Stampa…

2-Methylcinnamic acid ≥98.0% (by GC, titration analysis)

Fornitore: TCI
Attenzione

M1295-25G M1295-5G
TCIAM1295-25GEA 132 EUR
TCIAM1295-25G TCIAM1295-5G
2-Methylcinnamic acid ≥98.0% (by GC, titration analysis)
2-Methylcinnamic acid
Formula: C₁₀H₁₀O₂
Punto di ebollizione: 303 °C (1013 hPa)
Punto di fusione: 175…176 °C
MDL Number: MFCD00016841
CAS Number: 2373-76-4
EINECS: 219-150-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7a01f1849b5bc7b3c014d04a19275f17a7c9df8b&vid=7a01f1849b5bc7b3c014d04a19275f17a7c9df8b&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 175.0 to 178.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR