Stampa…

2,4,5-Trimethylbenzoic acid ≥97.0% (by GC, titration analysis)

Fornitore: TCI EUROPE

T2469-25G T2469-5G
TCIAT2469-25GEA 421 EUR
TCIAT2469-25G TCIAT2469-5G
2,4,5-Trimethylbenzoic acid ≥97.0% (by GC, titration analysis)
2,4,5-Trimethylbenzoic acid
Formula: C₁₀H₁₂O₂
Peso molecolare: 164,2 g/mol
MDL Number: MFCD00075863
CAS Number: 528-90-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4cc03026d9062f55c72dac3af2dbbd79805f2d6e&vid=4cc03026d9062f55c72dac3af2dbbd79805f2d6e&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 97.0 %
Purity (Neutralization titration) min. 97.0 %
Melting point 150.0 to 154.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR