Stampa…

D(-)-Glutammina ≥98.0% (by titrimetric analysis)

Fornitore: TCI

G0278-1G G0278-5G
TCIAG0278-1GEA 41 EUR
TCIAG0278-1G TCIAG0278-5G
D(-)-Glutammina ≥98.0% (by titrimetric analysis)
D(-)-Glutammina
Formula: C₅H₁₀N₂O₃
MDL Number: MFCD00065607
CAS Number: 5959-95-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f3f046fbc0f8ca30b81615db5c5a305ad0d16941&vid=f3f046fbc0f8ca30b81615db5c5a305ad0d16941&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Optical purity (LC) min. 98.0 ee%
Purity (Nonaqueous Titration) min. 98.0 %
Specific rotation [a]20/D -7.5 to -5.0 deg (C=4, H2O)
Solubility in Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR