Stampa…

Benzoyl fluoride ≥98.0% (by GC, titration analysis)

Fornitore: TCI
Pericolo

B1115-25G B1115-5G
TCIAB1115-25GEA 132 EUR
TCIAB1115-25G TCIAB1115-5G
Benzoyl fluoride ≥98.0% (by GC, titration analysis)
Benzoyl fluoride
Formula: C₇H₅FO
Punto di ebollizione: 160°C
Punto di fusione: –28,5 °C
Densità: 1,155 g/cm³ (18 °C)
Flash Pt: 49°C
MDL Number: MFCD00000364
CAS Number: 455-32-3
EINECS: 207-244-9
UN: 2920
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8072e4beafb0cb7b669ef6473c3fcb139aff35ba&vid=8072e4beafb0cb7b669ef6473c3fcb139aff35ba&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Brown clear liquid
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR