Stampa…

5-Methoxyisatin ≥95.0% (by HPLC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 5-Methoxy-1H-indole-2,3-dione, 5-Methoxyindoline-2,3-dione

M1362-25G M1362-5G
TCIAM1362-25GEA 162 EUR
TCIAM1362-25G TCIAM1362-5G
5-Methoxyisatin ≥95.0% (by HPLC, titration analysis)
5-Methoxyisatin
Formula: C₉H₇NO₃
Peso molecolare: 177,16 g/mol
MDL Number: MFCD00169023
CAS Number: 39755-95-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a116ffbd04bc1bc453cc9a1fbf1449574a40bde9&vid=a116ffbd04bc1bc453cc9a1fbf1449574a40bde9&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Orange to Amber to Dark red powder to crystal
Purity (HPLC) min. 95.0 area%
Purity (Neutralization titration) min. 95.0 %
Melting point 200.0 to 205.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR