2-Amino-5-chloro-3-nitropyridine ≥98.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
5-Chloro-3-nitro-2-pyridylamine, 5-Chloro-3-nitropyridin-2-amine
A1557-25G
A1557-5G
TCIAA1557-25GEA
89
EUR
TCIAA1557-25G
TCIAA1557-5G
2-Amino-5-chloro-3-nitropyridine ≥98.0% (by GC)
2-Amino-5-chloro-3-nitropyridine
Formula:
C₅H₄ClN₃O₂ Peso molecolare: 173,56 g/mol |
MDL Number:
MFCD00092011 CAS Number: 5409-39-2 |
Risultati del test di specificazione
Appearance | Light yellow to Brown to Dark green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 194.0 to 198.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...