Stampa…

2-Chloro-3-iodopyridine ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

C2193-1G C2193-5G
TCIAC2193-1GEA 16 EUR
TCIAC2193-1G TCIAC2193-5G
2-Chloro-3-iodopyridine ≥98.0% (by GC)
2-Chloro-3-iodopyridine
Formula: C₅H₃ClIN
Peso molecolare: 239,44 g/mol
MDL Number: MFCD00661298
CAS Number: 78607-36-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3480ec77c7bc490c5fcc9401213aecfb46161f9f&vid=3480ec77c7bc490c5fcc9401213aecfb46161f9f&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 93.0 to 97.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR