Chlorodifluoroacetic anhydride ≥98.0% (by titrimetric analysis)
Fornitore: TCI EUROPE
|
Pericolo
|
C1503-10G
C1503-25G
TCIAC1503-10GEA
80
EUR
TCIAC1503-10G
TCIAC1503-25G
Chlorodifluoroacetic anhydride ≥98.0% (by titrimetric analysis)
Chlorodifluoroacetic anhydride
Formula:
C₄Cl₂F₄O₃ Peso molecolare: 242,94 g/mol Punto di ebollizione: 93,5 - 94 °C (1013 hPa) Densità: 1,3953 g/cm³ (20 °C) |
MDL Number:
MFCD00000774 CAS Number: 2834-23-3 EINECS: 220-603-4 UN: 3265 ADR: 8,II |
Risultati del test di specificazione
Appearance | Colorless to Almost colorless clear liquid |
Purity (Neutralization titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...