Stampa…

2-Chloro-5-nitrocinnamic acid ≥97.0% (by titrimetric analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 3-(2-Chloro-5-nitrophenyl)acrylic acid

C1206-25G C1206-5G
TCIAC1206-25GEA 237 EUR
TCIAC1206-25G TCIAC1206-5G
2-Chloro-5-nitrocinnamic acid ≥97.0% (by titrimetric analysis)
2-Chloro-5-nitrocinnamic acid
Formula: C₉H₆ClNO₄
Peso molecolare: 227,6 g/mol
Punto di fusione: 223…234 °C
MDL Number: MFCD00063312
CAS Number: 36015-19-7
ADR: 9,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a648e50b9b42db8258adb527b0f6394655320cc3&vid=a648e50b9b42db8258adb527b0f6394655320cc3&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Yellow to Green powder to crystal
Purity (Neutralization titration) min. 97.0 %
Solubility in hot Methanol very faint turbidity

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR