2-Chloronicotinic acid ≥98.0% (by HPLC, titration analysis)
Fornitore: TCI
|
Attenzione
|
Synonyms:
2-Chloro-3-pyridinecarboxylic acid, 2-Chloropyridine-3-carboxylic acid
Formula:
C₆H₄ClNO₂ Punto di ebollizione: 284 °C (1013 hPa) Punto di fusione: 181 °C Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00006236 CAS Number: 2942-59-8 EINECS: 220-937-0 |
|
Risultati del test di specificazione
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Solubility in hot Methanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...