2-Chloronicotinamide ≥98.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
C1404-25G
C1404-5G
TCIAC1404-25GEA
89
EUR
TCIAC1404-25G
TCIAC1404-5G
2-Chloronicotinamide ≥98.0% (by GC)
2-Chloronicotinamide
Formula:
C₆H₅ClN₂O Peso molecolare: 156,57 g/mol Punto di fusione: 164…165 °C Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00006237 CAS Number: 10366-35-5 EINECS: 233-808-9 |
Risultati del test di specificazione
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 162.0 to 167.0 deg-C |
Solubility in hot Water | very faint turbidity |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...