Stampa…

Tetraphenylphosphonium chloride ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

T1375-25G T1375-5G
TCIAT1375-25GEA 81 EUR
TCIAT1375-25G TCIAT1375-5G
Tetraphenylphosphonium chloride ≥98.0% (by HPLC, titration analysis)
Tetraphenylphosphonium chloride
Formula: C₂₄H₂₀ClP
Peso molecolare: 374,85 g/mol
Punto di fusione: 275…278 °C
MDL Number: MFCD00011916
CAS Number: 2001-45-8
EINECS: 217-890-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4256807b4cad2fe46c1fa7adc002514337c31608&vid=4256807b4cad2fe46c1fa7adc002514337c31608&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Solubility in Water within almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR