Stampa…

3,4,5-Trihydroxybenzaldehyde ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: Gallic aldehyde, Pyrogallol-5-carboxaldehyde

T2650-1G T2650-5G
TCIAT2650-1GEA 111 EUR
TCIAT2650-1G TCIAT2650-5G
3,4,5-Trihydroxybenzaldehyde ≥98.0% (by GC, titration analysis)
3,4,5-Trihydroxybenzaldehyde
Formula: C₇H₆O₄
Peso molecolare: 154,12 g/mol
MDL Number: MFCD00003371
CAS Number: 13677-79-7
EINECS: 237-168-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ca14924719201197601ddcfb5924d6ed12c1c05e&vid=ca14924719201197601ddcfb5924d6ed12c1c05e&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light gray to Light orange powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR