Stampa…

trans-2-Undecenal ≥93.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: (E)-2-Undecenal

U0046-5ML U0046-25ML
TCIAU0046-5MLEA 18 EUR
TCIAU0046-5ML TCIAU0046-25ML
trans-2-Undecenal ≥93.0% (by GC)
trans-2-Undecenal
Formula: C₁₁H₂₀O
Peso molecolare: 168,28 g/mol
Flash Pt: 120°C
MDL Number: MFCD00014680
CAS Number: 53448-07-0
EINECS: 258-559-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7b5156b0c6e7c291ebf1106e421d184eb609f123&vid=7b5156b0c6e7c291ebf1106e421d184eb609f123&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 93.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR