4-Bromoanthranilic acid ≥98.0% (by HPLC, titration analysis)
Fornitore: TCI
|
Attenzione
|
Formula:
C₇H₆BrNO₂ |
MDL Number:
MFCD03618454 CAS Number: 20776-50-5 EINECS: 244-025-7 ADR: 6.1,III |
|
Risultati del test di specificazione
Appearance | White to Orange to Green powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...