Stampa…

2-Bromocinnamic acid ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

B1943-25G B1943-5G
TCIAB1943-25GEA 146 EUR
TCIAB1943-25G TCIAB1943-5G
2-Bromocinnamic acid ≥98.0% (by GC, titration analysis)
2-Bromocinnamic acid
Formula: C₉H₇BrO₂
MDL Number: MFCD00016836
CAS Number: 7345-79-1
EINECS: 231-353-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=264d8b50ad64deb09d75cf060eb027fa87967f49&vid=fadd16581c48ddecd8e1a51cc006605b7063a22f&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 216.0 to 220.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR