Stampa…

(2-Bromophenyl)acetic acid ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 2-(2-Bromophenyl)acetic acid

B1957-25G B1957-5G
TCIAB1957-25GEA 76 EUR
TCIAB1957-25G TCIAB1957-5G
(2-Bromophenyl)acetic acid ≥98.0% (by GC, titration analysis)
(2-Bromophenyl)acetic acid
Formula: (C₆H₄Br)CH₂COOH
Peso molecolare: 215,05 g/mol
Punto di ebollizione: 308 °C (1013 hPa)
Punto di fusione: 107,5 °C
MDL Number: MFCD00004314
CAS Number: 18698-97-0
EINECS: 242-509-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=02e1b5001b74bae93edcc3ba5757dd6946b1fae5&vid=02e1b5001b74bae93edcc3ba5757dd6946b1fae5&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 104.0 to 108.0 deg-C
Solubility in Methanol very faint turbidity

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR