ω-Bromoacetofenone ≥98.0% (by GC)
Fornitore: TCI EUROPE
|
Pericolo
|
Synonyms:
α-Bromoacetophenone, Phenacyl bromide, 2-Bromoacetophenone
B0535-100G
B0535-25G
TCIAB0535-100GEA
137
EUR
TCIAB0535-100G
TCIAB0535-25G
ω-Bromoacetofenone ≥98.0% (by GC)
ω-Bromoacetofenone
Formula:
(C₆H₅)C(O)CH₂Br Peso molecolare: 199,05 g/mol Punto di ebollizione: 135°C Punto di fusione: 50 °C Densità: 1,647 g/cm³ (20 °C) Flash Pt: ≥ 112 °C Temperatura di stoccaggio: Refrigerata |
MDL Number:
MFCD00000195 CAS Number: 70-11-1 EINECS: 200-724-9 UN: 2645 ADR: 6.1,II Merck Index: 14,01402 |
Risultati del test di specificazione
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 49.0 to 52.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...