4-Bromo-2-chlorobenzotrifluoride ≥98.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
B3057-25G
B3057-5G
TCIAB3057-25GEA
106
EUR
TCIAB3057-25G
TCIAB3057-5G
4-Bromo-2-chlorobenzotrifluoride ≥98.0% (by GC)
4-Bromo-2-chlorobenzotrifluoride
Formula:
C₇H₃BrClF₃ Peso molecolare: 259,45 g/mol |
MDL Number:
MFCD08459292 CAS Number: 467435-07-0 |
Risultati del test di specificazione
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...