3-Bromo-4,5-dimethoxybenzaldehyde ≥98.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
5-Bromoveratraldehyde
B3130-25G
B3130-5G
TCIAB3130-25GEA
459
EUR
TCIAB3130-25G
TCIAB3130-5G
3-Bromo-4,5-dimethoxybenzaldehyde ≥98.0% (by GC)
3-Bromo-4,5-dimethoxybenzaldehyde
Formula:
C₉H₉BrO₃ Peso molecolare: 245,07 g/mol |
MDL Number:
MFCD00003346 CAS Number: 6948-30-7 EINECS: 230-113-2 |
Risultati del test di specificazione
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 60.0 to 64.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...