Stampa…

Benzyltriethylammonium borhydride ≥90.0% (by titrimetric analysis)

Fornitore: TCI EUROPE
Pericolo

Synonyms: Benzyltriethylammonium Borohydride

B3128-25G B3128-5G
TCIAB3128-25GEA 315 EUR
TCIAB3128-25G TCIAB3128-5G
Benzyltriethylammonium borhydride ≥90.0% (by titrimetric analysis)
Benzyltriethylammonium borhydride
Formula: C₁₃H₂₆BN
Peso molecolare: 207,17 g/mol
Punto di fusione: 143°C
MDL Number: MFCD00191785
CAS Number: 85874-45-9
UN: 3131
ADR: 4.3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=725a1556adcbe3b24cece810dba76f13b249da61&vid=725a1556adcbe3b24cece810dba76f13b249da61&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (Sodium hypochlorite Method) min. 90.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR