Stampa…

2,4,6-Trimethylphenylboronic acid ≥98.0% (by HPLC)

Fornitore: TCI
Attenzione

T1960-1G T1960-5G
TCIAT1960-1GEA 28 EUR
TCIAT1960-1G TCIAT1960-5G
2,4,6-Trimethylphenylboronic acid ≥98.0% (by HPLC)
2,4,6-Trimethylphenylboronic acid
Formula: C₉H₁₃BO₂
MDL Number: MFCD00236060
CAS Number: 5980-97-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=521e665a6d2dd56c7b1eb83598ec91dbc50aa0b7&vid=521e665a6d2dd56c7b1eb83598ec91dbc50aa0b7&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR