Stampa…

Diphenyleneiodonium chloride

Fornitore: Cayman Chemical

Synonyms: Diphenyleneiodonium Chloride

81050-10 81050-5
CAYM81050-10EA 152 EUR
CAYM81050-10 CAYM81050-5
Diphenyleneiodonium chloride
Diphenyleneiodonium chloride
Formula: C₁₂H₈ClI
Peso molecolare: 314,55 g/mol
MDL Number: MFCD00214165
CAS Number: 4673-26-1

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR