Stampa…

(S)-(-)-α,α-Diphenylprolinol 99+%

Fornitore: Thermo Scientific
Attenzione

Synonyms: (S)-(-)-α,α-Diphenyl-2-pyrrolidinemethanol

290420010 290420050
ACRO290420010EA 54.4 EUR
ACRO290420010 ACRO290420050
(S)-(-)-α,α-Diphenylprolinol 99+%
(S)-(-)-α,α-Diphenylprolinol
Formula: C₁₇H₁₉NO
Peso molecolare: 253,35 g/mol
Punto di fusione: 77…80 °C
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00075506
CAS Number: 112068-01-6

Visualizza i codici

Risultati del test di specificazione

Appearance White to beige Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 77 to 80 °C
GC ≥99.0 %
Specific optical rotation -57° to -61° (25 °C, 589 nm) (c=3, MeOH)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR