Stampa…
(±)-11-HDoHE
Fornitore: Cayman Chemical
33450-25 33450-50 33450-100 33450-250
CAYM33450-25EA 168 EUR
CAYM33450-25 CAYM33450-50 CAYM33450-100 CAYM33450-250
(±)-11-HDoHE
(±)-11-HDoHE
Formula: C₂₂H₃₂O₃
CAS Number: 87018-59-5
UN: 1170
ADR: 3,II