Stampa…

5-Acetylthiophene-2-boronic acid (contains varying amounts of Anhydride)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 5-Acetyl-2-thiopheneboronic acid

A2364-25G A2364-5G
TCIAA2364-25GEA 595 EUR
TCIAA2364-25G TCIAA2364-5G
5-Acetylthiophene-2-boronic acid (contains varying amounts of Anhydride)
5-Acetylthiophene-2-boronic acid
Formula: C₆H₇BO₃S
Peso molecolare: 170,00 g/mol
MDL Number: MFCD01075681
CAS Number: 206551-43-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2ed15abe0781b4ac9f690c6b2a408c8096045de8&vid=2ed15abe0781b4ac9f690c6b2a408c8096045de8&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Gray to Brown powder to crystal
Purity (Neutralization titration) 97.0 to 112.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR