Stampa…

3-Hydroxypyridine-2-carbonitrile ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Pericolo

Synonyms: 2-Cyano-3-hydroxypyridine, 3-Hydroxypicolinonitrile

C1664-25G C1664-5G
TCIAC1664-25GEA 651 EUR
TCIAC1664-25G TCIAC1664-5G
3-Hydroxypyridine-2-carbonitrile ≥98.0% (by GC, titration analysis)
3-Hydroxypyridine-2-carbonitrile
Formula: C₆H₄N₂O
Peso molecolare: 120,1088 g/mol
MDL Number: MFCD01646135
CAS Number: 932-35-4
EINECS: 213-251-8
UN: 3439
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=842c117014453420dd38353edd5c5441c01a7fcd&vid=842c117014453420dd38353edd5c5441c01a7fcd&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR