Stampa…

Triphenylmethyl mercaptan ≥98%

Fornitore: Thermo Scientific
Attenzione

Synonyms: Triphenylmethanethiol

A14888.06 A14888.14 A14888.22
A14888.06EA 32.8 EUR
A14888.06 A14888.14 A14888.22
Triphenylmethyl mercaptan ≥98%
Triphenylmethyl mercaptan

Purity: 98+% CAS No.: 3695-77-0 Molecular Formula: (C6H5)3CSH Formula Weight: 276.40 Odour: Stench Melting Point: 103-108° Beilstein Reference: 2113168 MDL No.: MFCD00004854

Formula: C₁₉H₁₆S
Peso molecolare: 276,40 g/mol
Punto di fusione: 103…108 °C
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00004854
CAS Number: 3695-77-0
EINECS: 223-020-3

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR