Tetrabromobisphenol A ≥98.0% (by GC, titration analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
4,4'-(Propane-2,2-diyl)bis(2,6-dibromophenol), 3,3',5,5'-Tetrabromobisphenol A
T0032-25G
T0032-500G
TCIAT0032-25GEA
18
EUR
TCIAT0032-25G
TCIAT0032-500G
Tetrabromobisphenol A ≥98.0% (by GC, titration analysis)
Tetrabromobisphenol A
Formula:
C₁₅H₁₂Br₄O₂ Peso molecolare: 543,87 g/mol |
MDL Number:
MFCD00013962 CAS Number: 79-94-7 EINECS: 201-236-9 UN: 3077 ADR: 9,III |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 178.0 to 184.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...