Stampa…
2-Chloroethanesulphonic acid sodium salt hydrate, (max. 10% H₂O) 98+% (dry weight)
Fornitore: Thermo Fisher Scientific
Attenzione
Synonyms: Sodium-2-chloroethanesulphonate hydrate
A19995.06 A19995.14 A19995.22
A19995.06EA 27.7 EUR
A19995.06 A19995.14 A19995.22
2-Chloroethanesulphonic acid sodium salt hydrate, (max. 10% H₂O) 98+% (dry weight)
2-Chloroethanesulphonic acid sodium salt hydrate
Purity: 98+% CAS No.: 15484-44-3 Molecular Formula: Cl(CH2)2SO3Na.xH2O Formula Weight: 166.56(anhy) Melting Point: ca 292° dec. Beilstein Reference: 5649202
Formula: C₂H₅ClO₃S·H₂O
Peso molecolare: 184.57 g/mol
Punto di fusione: ∼ 292 °C (dec.)
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00007531
CAS Number: 15484-44-3
EINECS: 239-508-4