Stampa…

Tetraclorometano-[13C] (99% ¹³C )

Fornitore: LGC Standards PROMOCHEM
Pericolo

CILCLM-731-0.1 CILCLM-731-0.5 CIL-CLM-731-1
PROOCILCLM-731-0.1EA 125 EUR
PROOCILCLM-731-0.1 PROOCILCLM-731-0.5 PROOCIL-CLM-731-1
Tetraclorometano-[13C] (99% ¹³C )
Tetraclorometano-[13C]
Formula: CCl₄
Temperatura di stoccaggio: Ambiente
CAS Number: 32488-50-9
UN: 1846
ADR: 6.1,II

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR