Cyclopropylacetic acid ≥97%
Fornitore: Thermo Fisher Scientific
|
Pericolo
|
Purity: 97%
CAS No.: 5239-82-7
Molecular Formula: C5H8O2
Formula Weight: 100.12
Flash Point: 93°(199°F)
Refractive Index: 1.4350
Odour: Pungent
Boiling Point: 189-191°
Beilstein Reference: 2323311
UN No.: 3265
MDL No.: MFCD00041544
Formula:
C₅H₈O₂ Punto di ebollizione: 189…191 °C Densità: 1,076 Flash Pt: 93 °C (199 °F) Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00041544 CAS Number: 5239-82-7 UN: 3265 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...