2-Amino-5-bromopyrazine 97%
Fornitore: Thermo Fisher Scientific
|
Pericolo
|
Synonyms:
5-bromopyrazin-2-amine
Purity: 97%
CAS No.: 59489-71-3
Molecular Formula: C4H4BrN3
Formula Weight: 174.00
Melting Point: 113-117°
MDL No.: MFCD00235015
Formula:
C₄H₄BrN₃ Peso molecolare: 174.00 g/mol Punto di fusione: 113…117 °C Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00235015 CAS Number: 59489-71-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...