Phenylglyoxylic acid 97%
Fornitore: Thermo Fisher Scientific
|
Attenzione
|
Synonyms:
2-Oxo-2-phenylacetic acid, Benzoylformic acid
Formula:
C₈H₆O₃ Peso molecolare: 150.13 g/mol Punto di fusione: 60…69 °C Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00002575 CAS Number: 611-73-4 EINECS: 210-278-7 |
Risultati del test di specificazione
Appearance | White to light yellow Crystalline powder or crystals and/or chunks |
Infrared spectrum | Conforms |
Melting point | 60 to 69 °C |
Titration with NaOH | ≥96.0 % |
GC | ≥98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...